Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC37A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164277.1 | NP_001157749.1 | 4 | 940 | ||
NM_001164278.1 | NP_001157750.1 | 4 | 624 | |||
NM_001164279.1 | NP_001157751.1 | 4 | 366 | |||
NM_001164280.1 | NP_001157752.1 | 2 | 382 | |||
NM_001467.5 | NP_001458.1 | 3 | 436 | |||
GenBank mRNA | AB209026.1 | - | 1 | 2472 | ||
AF110819.1 | - | 2 | 183 | |||
AF110820.1 | - | 2 | 183 | |||
AF110821.1 | - | 2 | 183 | |||
AF110822.1 | - | 2 | 183 | |||
AF111852.1 | - | 1 | 615 | |||
AK289642.1 | - | 4 | 908 | |||
AK294993.1 | - | 4 | 502 | |||
AK298395.1 | - | 4 | 320 | |||
AK300646.1 | - | 4 | 332 | |||
AK312628.1 | - | 4 | 606 | |||
AY423732.1 | - | 2 | 203 | |||
BC002400.2 | - | 2 | 384 | 2822215 | ||
BC003589.1 | - | 2 | 384 | 2822215 | ||
BC014663.1 | - | 2 | 391 | 4857067 | ||
BC015650.2 | - | 3 | 856 | 4660508 | ||
BC064563.1 | - | 2 | 379 | 5580134 | ||
CR456733.1 | - | 2 | 183 | |||
DA485567.1 | - | 3 | 410 | |||
DC295438.1 | - | 3 | 436 | |||
Y15409.1 | - | 2 | 352 |