Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002872.4 | NP_002863.1 | 5, 6 | 600 | ||
GenBank mRNA | AF498965.1 | - | 5, 6 | 449 | ||
BC001485.2 | - | 5, 6 | 572 | 3509104 | ||
BC018735.1 | - | 1, 2 | 119 | |||
BI908751.1 | - | 5, 6 | 602 | |||
BQ934070.1 | - | 5, 6 | 568 | |||
BT006919.1 | - | 5, 6 | 449 | |||
CR456555.1 | - | 5, 6 | 482 | |||
M29871.1 | - | 5, 6 | 449 |