Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297437.1 | NP_001284366.1 | 5 | 824 | ||
NM_012176.2 | NP_036308.1 | 5 | 822 | |||
NM_033484.2 | NP_277019.1 | 5 | 822 | |||
XM_006714470.3 | XP_006714533.1 | 4 | 520 | |||
XM_011514026.2 | XP_011512328.1 | 5 | 827 | |||
XM_011514027.2 | XP_011512329.1 | 5 | 2197 | |||
GenBank mRNA | AF129534.1 | - | 5 | 766 | ||
AF176703.1 | - | 5 | 760 | |||
AK024075.1 | - | 5 | 822 | |||
BC040011.1 | - | 5 | 783 | |||
BC048098.1 | - | 5 | 790 | 4830192 | ||
CR749719.1 | - | 3 | 339 |