Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC61A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013336.3 | NP_037468.1 | 10 | 1276 | ||
GenBank mRNA | AF077032.1 | - | 10 | 1182 | ||
AF084458.1 | - | 10 | 1182 | |||
AF346602.1 | - | 10 | 1158 | |||
AK001689.1 | - | 3 | 336 | |||
AK027423.1 | - | 10 | 1276 | |||
AK027452.1 | - | 2 | 242 | |||
AK027454.1 | - | 1 | 62 | |||
AK027458.1 | - | 1 | 21 | |||
AK074706.1 | - | 10 | 1276 | |||
AK074907.1 | - | 2 | 246 | |||
AK074928.1 | - | 6 | 884 | |||
AK075148.1 | - | 2 | 276 | |||
AK299118.1 | - | 10 | 1356 | |||
AL831940.1 | - | 5 | 686 | |||
AL832821.1 | - | 10 | 1209 | |||
BC002951.1 | - | 10 | 1183 |