Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014564.4 | NP_055379.1 | 2, 3 | 394 | ||
NM_178138.5 | NP_835258.1 | 2, 3 | 362 | |||
XM_005263410.1 | XP_005263467.1 | 2, 3 | 676 | |||
XM_017015168.1 | XP_016870657.1 | 1, 2 | 173 | |||
GenBank mRNA | AB593062.1 | - | 2, 3 | 362 | ||
AB593063.1 | - | 2, 3 | 361 | |||
AB593064.1 | - | 2, 3 | 676 | |||
AB593066.1 | - | 2, 3 | 394 | |||
AB593067.1 | - | 2, 3 | 358 | |||
AF096169.1 | - | 1, 2 | 166 | |||
AF156888.1 | - | 2, 3 | 349 | |||
AF156889.1 | - | 2, 3 | 379 |