Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIKESHI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322407.1 | NP_001309336.1 | 5 | 836 | ||
NM_001322409.1 | NP_001309338.1 | 5 | 766 | |||
NM_016401.3 | NP_057485.2 | 4 | 678 | |||
NR_024597.1 | 3 | 440 | ||||
NR_024598.1 | 3 | 440 | ||||
NR_136324.1 | 5 | 786 | ||||
XM_017017914.1 | XP_016873403.1 | 4 | 668 | |||
XM_017017915.1 | XP_016873404.1 | 4 | 455 | |||
GenBank mRNA | AA148558.1 | - | 3 | 405 | ||
AF151082.1 | - | 3 | 321 | |||
AF161487.1 | - | 4 | 659 | |||
AF161527.1 | - | 4 | 662 | |||
AK001447.1 | - | 4 | 649 | |||
AK125010.1 | - | 1 | 2356 | |||
AK223198.1 | - | 4 | 649 | |||
AK298763.1 | - | 5 | 802 | |||
BC001677.2 | - | 4 | 551 | 2820772 | ||
BC006476.2 | - | 4 | 551 | 2820772 | ||
BC015991.2 | - | 4 | 643 | 2820691 | ||
BC018080.1 | - | 4 | 612 | 4798958 | ||
BC021621.1 | - | 4 | 643 | 2820691 | ||
BP423222.1 | - | 3 | 357 | |||
BX332668.2 | - | 4 | 515 |