Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131008.1 | NP_001124480.1 | 4 | 303 | ||
NM_002835.3 | NP_002826.3 | 4 | 421 | |||
XM_011516444.1 | XP_011514746.1 | 4 | 388 | |||
GenBank mRNA | AB209524.1 | - | 4 | 412 | ||
AK289573.1 | - | 4 | 365 | |||
AK303602.1 | - | 4 | 303 | |||
AK314236.1 | - | 4 | 388 | |||
BC050008.1 | - | 5 | 606 | 5268732 | ||
CN395133.1 | - | 4 | 424 | |||
D13380.1 | - | 4 | 349 | |||
M93425.1 | - | 4 | 359 | |||
S69182.1 | - | 2 | 79 | |||
S69184.1 | - | 3 | 156 |