Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122770.1 | NP_001116242.1 | 3 | 368 | ||
NM_032522.3 | NP_115911.1 | 3 | 365 | |||
XM_005245546.2 | XP_005245603.1 | 3 | 423 | |||
XM_006711578.3 | XP_006711641.1 | 2 | 432 | |||
XM_011510062.2 | XP_011508364.1 | 3 | 423 | |||
XM_017002556.1 | XP_016858045.1 | 2 | 433 | |||
XM_017002557.1 | XP_016858046.1 | 3 | 343 | |||
XM_017002558.1 | XP_016858047.1 | 3 | 423 | |||
GenBank mRNA | AF318371.1 | - | 2 | 284 | ||
AK057310.1 | - | 3 | 491 | |||
AK302696.1 | - | 3 | 340 | |||
BC003116.1 | - | 2 | 326 | 3503643 | ||
BC006315.2 | - | 2 | 326 | 4124255 | ||
DA790498.1 | - | 3 | 343 | |||
DA796002.1 | - | 3 | 365 |