Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242597.1 | NP_001229526.1 | 1 | 337 | ||
NM_001242598.1 | NP_001229527.1 | 2 | 266 | |||
NM_032635.3 | NP_116024.1 | 2 | 266 | |||
GenBank mRNA | BC001118.2 | - | 2 | 223 | 2989840 | |
BG697070.1 | - | 1 | 226 | |||
BM423809.1 | - | 2 | 266 | |||
BM765457.1 | - | 2 | 76 | |||
Y18007.1 | - | 2 | 234 |