Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DISP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032890.3 | NP_116279.2 | 4 | 448 | ||
XM_005273335.2 | XP_005273392.1 | 2 | 289 | |||
XM_006711592.2 | XP_006711655.1 | 3 | 1236 | |||
XM_011510072.2 | XP_011508374.1 | 3 | 288 | |||
XM_011510073.2 | XP_011508375.1 | 3 | 766 | |||
XM_011510074.2 | XP_011508376.1 | 3 | 307 | |||
XM_011510075.2 | XP_011508377.1 | 3 | 794 | |||
XM_011510077.2 | XP_011508379.1 | 2 | 230 | |||
XM_017002611.1 | XP_016858100.1 | 2 | 704 | |||
GenBank mRNA | AK098669.1 | - | 2 | 182 | ||
AK125728.1 | - | 2 | 286 | |||
BC007734.2 | - | 2 | 269 | |||
BC107762.1 | - | 2 | 273 | |||
DA756409.1 | - | 4 | 448 |