Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011655.2 | NP_001011655.1 | 7 | 1085 | ||
NM_001166305.1 | NP_001159777.1 | 8 | 1226 | |||
NM_001166306.1 | NP_001159778.1 | 7 | 1085 | |||
NM_138399.4 | NP_612408.3 | 7 | 1085 | |||
XM_005269371.4 | XP_005269428.1 | 7 | 1099 | |||
XM_011513318.2 | XP_011511620.1 | 8 | 1129 | |||
XM_011513319.2 | XP_011511621.1 | 7 | 1020 | |||
XM_011513320.2 | XP_011511622.1 | 8 | 1250 | |||
XM_011513321.2 | XP_011511623.1 | 7 | 996 | |||
XM_011513322.2 | XP_011511624.1 | 7 | 1106 | |||
XM_017007517.1 | XP_016863006.1 | 7 | 987 | |||
XM_017007518.1 | XP_016863007.1 | 7 | 982 | |||
GenBank mRNA | AK126914.1 | - | 8 | 882 | ||
AL833026.1 | - | 2 | 643 | |||
AY303778.1 | - | 1 | 75 | |||
AY597813.1 | - | 1 | 75 | |||
BC007772.2 | - | 2 | 231 | |||
BC014883.1 | - | 7 | 945 | 3907191 | ||
BC034353.1 | - | 2 | 153 | |||
BC111485.1 | - | 8 | 1185 | 5767916 | ||
BC130297.1 | - | 7 | 989 | 40146328 | ||
BC144158.1 | - | 7 | 989 | 9052678 | ||
BC144159.1 | - | 7 | 989 | 9052679 | ||
BX647418.1 | - | 7 | 844 | |||
CF265142.1 | - | 2 | 240 |