Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138423.3 | NP_612432.2 | 1 | 636 | ||
NM_177974.2 | NP_816929.1 | 1 | 636 | |||
XM_017021880.1 | XP_016877369.1 | 1 | 731 | |||
GenBank mRNA | AK098265.1 | - | 1 | 609 | ||
AK291026.1 | - | 1 | 393 | |||
AK298564.1 | - | 2 | 351 | |||
AL832333.1 | - | 1 | 408 | |||
AL832840.1 | - | 1 | 78 | |||
AY358086.1 | - | 1 | 587 | |||
BC012124.1 | - | 1 | 616 | |||
BC063480.1 | - | 1 | 590 | 5218924 | ||
BG327802.1 | - | 615 |