Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACYP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320586.1 | NP_001307515.1 | 5, 6 | 553 | ||
NM_001320587.1 | NP_001307516.1 | 4, 5 | 460 | |||
NM_001320588.1 | NP_001307517.1 | 2, 3 | 254 | |||
NM_001320589.1 | NP_001307518.1 | 2, 3 | 505 | |||
NM_001320590.1 | NP_001307519.1 | 2, 3 | 505 | |||
NM_138448.3 | NP_612457.1 | 2, 3 | 505 | |||
XM_017005406.1 | XP_016860895.1 | 6, 7 | 899 | |||
XM_017005407.1 | XP_016860896.1 | 5, 6 | 818 | |||
XM_017005408.1 | XP_016860897.1 | 4, 5 | 1527 | |||
XM_017005409.1 | XP_016860898.1 | 4, 5 | 725 | |||
XM_017005410.1 | XP_016860899.1 | 2, 3 | 375 | |||
XM_017005411.1 | XP_016860900.1 | 6, 7 | 899 | |||
XM_017005412.1 | XP_016860901.1 | 6, 7 | 899 | |||
XM_017005413.1 | XP_016860902.1 | 6, 7 | 899 | |||
GenBank mRNA | AK307096.1 | - | 2, 3 | 376 | ||
BC012290.1 | - | 2, 3 | 274 | 3864319 | ||
BF671963.1 | - | 2, 3 | 253 | |||
BG741807.1 | - | 4, 5 | 454 | |||
BU543943.1 | - | 5, 6 | 553 | |||
BU857820.1 | - | 2, 3 | 363 | |||
X84195.1 | - | 2, 3 | 254 |