Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204504.2 | NP_001191433.1 | 18 | 2230 | ||
NM_001204505.2 | NP_001191434.1 | 15 | 1851 | |||
NM_001204506.2 | NP_001191435.1 | 15 | 1851 | |||
NM_001297429.1 | NP_001284358.1 | 13 | 1618 | |||
NM_152910.5 | NP_690874.2 | 17 | 2193 | |||
NM_178009.4 | NP_821077.1 | 17 | 2193 | |||
NR_123714.1 | 15 | 1851 | ||||
NR_123715.1 | 17 | 2464 | ||||
GenBank mRNA | AB078967.1 | - | 17 | 2148 | ||
AB078968.1 | - | 17 | 2148 | |||
AK098302.1 | - | 2 | 212 | |||
AK122909.1 | - | 15 | 1816 | |||
AK289546.1 | - | 18 | 2232 | |||
AK302631.1 | - | 13 | 1578 | |||
AK302727.1 | - | 15 | 1851 | |||
AL833883.1 | - | 3 | 281 | |||
BC043292.2 | - | 15 | 1808 | 5297895 |