Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST7L SIRNA Assays ›
Gene Symbol:
ST7L
Gene Name:
suppression of tumorigenicity 7 like
Gene Aliases:
FAM4B, ST7R, STLR
Chromosome Location:
Chr.1: 112517444 - 112620838 on Build GRCh38
UniGene ID:
Hs.201921
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
43682
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST7L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308264.1
NP_001295193.1
3
474
NM_017744.4
NP_060214.2
3
628
NM_138727.3
NP_620055.1
3
577
NM_138728.2
NP_620056.1
3
628
NM_138729.3
NP_620057.1
3
628
XM_005270963.4
XP_005271020.1
3
151
XM_005270964.3
XP_005271021.1
2
243
XM_011541627.2
XP_011539929.1
3
337
XM_011541628.2
XP_011539930.1
3
337
XM_011541629.2
XP_011539931.1
3
351
XM_011541630.2
XP_011539932.1
3
1047
XM_011541631.2
XP_011539933.1
3
175
XM_017001534.1
XP_016857023.1
3
337
XM_017001535.1
XP_016857024.1
3
337
XM_017001536.1
XP_016857025.1
2
243
XM_017001537.1
XP_016857026.1
2
118
XM_017001538.1
XP_016857027.1
2
118
GenBank mRNA
AB081317.1
-
3
399
AB081318.1
-
3
348
AB081319.1
-
3
399
AB081320.1
-
3
399
AK021719.1
-
2
240
AK097837.1
-
3
263
AK291042.1
-
3
329
AK295507.1
-
3
326
AK295955.1
-
3
326
AK301869.1
-
3
329
AK304004.1
-
3
286
AK308727.1
-
2
252
BC027898.1
-
2
230
BC037830.1
-
3
1041
4812133
BC047392.1
-
3
354
5273549
BC065501.1
-
3
535
4417786
BX537988.1
-
4
490
BX640667.1
-
3
304
CD358134.1
-
3
628
Back To Top
More Information
Gene Ontology Categories:
Function(s)
negative regulation of cell growth
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-jlg4l:80/100.66.77.222:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline