Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206846.1 | NP_001193775.1 | 1 | 484 | ||
NM_018136.4 | NP_060606.3 | 1 | 484 | |||
GenBank mRNA | AF509326.1 | - | 1 | 227 | ||
AK125107.1 | - | 1 | 484 | |||
AK226178.1 | - | 1 | 417 | |||
AY099892.1 | - | 1 | 227 | |||
AY367065.1 | - | 1 | 227 | |||
AY971955.1 | - | 1 | 227 | |||
AY971956.1 | - | 1 | 227 | |||
AY971957.1 | - | 1 | 227 | |||
BC015396.2 | - | 1 | 325 |