Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011655.2 | NP_001011655.1 | 7, 8 | 1126 | ||
NM_001166305.1 | NP_001159777.1 | 8, 9 | 1267 | |||
NM_001166306.1 | NP_001159778.1 | 7, 8 | 1126 | |||
NM_138399.4 | NP_612408.3 | 7, 8 | 1126 | |||
XM_005269371.4 | XP_005269428.1 | 7, 8 | 1140 | |||
XM_011513318.2 | XP_011511620.1 | 8, 9 | 1170 | |||
XM_017007517.1 | XP_016863006.1 | 7, 8 | 1028 | |||
XM_017007518.1 | XP_016863007.1 | 7, 8 | 1023 | |||
GenBank mRNA | AK126914.1 | - | 8, 9 | 923 | ||
AL833026.1 | - | 2, 3 | 684 | |||
AY303778.1 | - | 1, 2 | 116 | |||
AY597813.1 | - | 1, 2 | 116 | |||
BC007772.2 | - | 2, 3 | 272 | |||
BC014883.1 | - | 7, 8 | 986 | 3907191 | ||
BC034353.1 | - | 2, 3 | 194 | |||
BC111485.1 | - | 8, 9 | 1226 | 5767916 | ||
BC130297.1 | - | 7, 8 | 1030 | 40146328 | ||
BC144158.1 | - | 7, 8 | 1030 | 9052678 | ||
BC144159.1 | - | 7, 8 | 1030 | 9052679 | ||
BX647418.1 | - | 7, 8 | 885 | |||
CF265142.1 | - | 2, 3 | 281 |