Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243266.1 | NP_001230195.1 | 2 | 323 | ||
NM_024021.3 | NP_076926.2 | 3 | 469 | |||
NM_148975.2 | NP_683876.1 | 2 | 323 | |||
XM_017017909.1 | XP_016873398.1 | 2 | 336 | |||
GenBank mRNA | AB013102.1 | - | 2 | 164 | ||
AB022821.1 | - | 2 | 188 | |||
AF068288.1 | - | 2 | 257 | |||
AF237912.1 | - | 2 | 274 | |||
AF350500.1 | - | 1 | 227 | |||
AF354928.1 | - | 1 | 147 | |||
AK075081.1 | - | 3 | 354 | |||
AK292089.1 | - | 2 | 278 | |||
AK314033.1 | - | 2 | 166 | |||
BC020648.1 | - | 2 | 198 | 4720088 | ||
BG538851.1 | - | 2 | 323 | |||
CB046393.1 | - | 2 | 192 |