Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207015.1 | NP_001193944.1 | 5 | 657 | ||
NM_001207016.1 | NP_001193945.1 | 3 | 447 | |||
NM_002849.3 | NP_002840.2 | 6 | 1313 | |||
NM_130846.2 | NP_570897.2 | 2 | 579 | |||
NR_073474.1 | 2 | 579 | ||||
XM_011538615.2 | XP_011536917.1 | 6 | 909 | |||
GenBank mRNA | AK295951.1 | - | 5 | 647 | ||
AK304457.1 | - | 3 | 406 | |||
AK304672.1 | - | 3 | 447 | |||
AK312376.1 | - | 6 | 1226 | |||
BC092469.1 | - | 2 | 579 | |||
BC110900.1 | - | 2 | 276 | 4538615 | ||
BC119777.1 | - | 2 | 255 | 40109494 | ||
BC122866.1 | - | 2 | 255 | 40109492 | ||
BX571751.1 | - | 2 | 632 | |||
D64053.1 | - | 6 | 1315 | |||
DC421975.1 | - | 3 | 447 | |||
U42361.1 | - | 6 | 1073 | |||
U77916.1 | - | 3 | 297 | |||
U77917.1 | - | 2 | 556 | |||
X82635.1 | - | 2 | 537 | |||
Z79693.1 | - | 6 | 984 |