Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018390.3 | NP_060860.1 | 6 | 911 | ||
NR_028057.1 | 7 | 894 | ||||
XM_006724865.2 | XP_006724928.1 | 7 | 852 | |||
XM_006724866.2 | XP_006724929.1 | 6 | 663 | |||
XM_006724867.2 | XP_006724930.1 | 6 | 684 | |||
XM_011545633.2 | XP_011543935.1 | 7 | 886 | |||
GenBank mRNA | AK002185.1 | - | 7 | 896 | ||
BC000888.1 | - | 6 | 817 | |||
BC005028.1 | - | 6 | 843 | 3953383 | ||
BC032072.1 | - | 6 | 817 |