Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256252.1 | NP_001243181.1 | 7, 8 | 862 | ||
NM_001256253.1 | NP_001243182.1 | 8, 9 | 1018 | |||
NM_001256254.1 | NP_001243183.1 | 5, 6 | 595 | |||
NM_001974.4 | NP_001965.3 | 8, 9 | 1018 | |||
XM_011527794.1 | XP_011526096.1 | 10, 11 | 1294 | |||
GenBank mRNA | AK131562.1 | - | 7, 8 | 832 | ||
AK290401.1 | - | 8, 9 | 988 | |||
AK291518.1 | - | 8, 9 | 988 | |||
AK297003.1 | - | 5, 6 | 573 | |||
AK313495.1 | - | 8, 9 | 988 | |||
BC059395.1 | - | 8, 9 | 1001 | 30349201 | ||
X81479.1 | - | 8, 9 | 996 |
Set Membership: |
G-protein coupled receptor |