Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021237.3 | NP_067060.2 | 4 | 400 | ||
GenBank mRNA | AF085359.1 | - | 4 | 356 | ||
AF161415.1 | - | 4 | 389 | |||
AF537132.1 | - | 4 | 292 | |||
AK289594.1 | - | 4 | 293 | |||
BC013162.1 | - | 4 | 296 | 4345594 | ||
BM762385.1 | - | 4 | 426 |