Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMAN2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142292.1 | NP_001135764.1 | 2 | 312 | ||
NM_001322346.1 | NP_001309275.1 | 2 | 328 | |||
NM_001322352.1 | NP_001309281.1 | 2 | 328 | |||
NM_001322354.1 | NP_001309283.1 | 2 | 342 | |||
NM_001322355.1 | NP_001309284.1 | 2 | 328 | |||
NM_001322356.1 | NP_001309285.1 | 2 | 328 | |||
NM_030805.3 | NP_110432.1 | 2 | 312 | |||
GenBank mRNA | AJ549957.1 | - | 2 | 276 | ||
AK075347.1 | - | 2 | 300 | |||
AK222828.1 | - | 2 | 290 | |||
AK299740.1 | - | 2 | 300 | |||
AK308085.1 | - | 2 | 299 | |||
AK310100.1 | - | 2 | 329 | |||
AK312249.1 | - | 2 | 305 | |||
AK316331.1 | - | 2 | 300 | |||
AL136617.1 | - | 2 | 304 | |||
AY358929.1 | - | 2 | 283 | |||
BC000347.2 | - | 2 | 275 | |||
BC005822.2 | - | 2 | 275 | |||
BC005862.2 | - | 2 | 275 | |||
BC067265.1 | - | 2 | 282 | 6502212 | ||
BX648002.1 | - | 2 | 290 | |||
DA521101.1 | - | 2 | 289 | |||
DC354352.1 | - | 2 | 314 |