Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RNF146 SIRNA Assays ›
Gene Symbol:
RNF146
Gene Name:
ring finger protein 146
Gene Aliases:
-
Chromosome Location:
Chr.6: 127266340 - 127289893 on Build GRCh38
UniGene ID:
Hs.267120
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
43779
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RNF146
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242844.1
NP_001229773.1
5
1004
NM_001242845.1
NP_001229774.1
5
853
NM_001242846.1
NP_001229775.1
5
766
NM_001242847.1
NP_001229776.1
4
895
NM_001242848.1
NP_001229777.1
4
625
NM_001242849.1
NP_001229778.1
3
767
NM_001242850.1
NP_001229779.1
3
497
NM_001242851.1
NP_001229780.1
3
437
NM_001242852.1
NP_001229781.1
5
984
NM_030963.3
NP_112225.2
5
734
XM_006715571.3
XP_006715634.1
5
1434
XM_011536161.2
XP_011534463.1
6
1321
XM_011536162.2
XP_011534464.1
6
1373
XM_011536163.2
XP_011534465.1
6
1353
XM_011536164.2
XP_011534466.1
5
1212
XM_017011336.1
XP_016866825.1
3
1114
XM_017011337.1
XP_016866826.1
5
4355
XM_017011338.1
XP_016866827.1
7
1462
XM_017011339.1
XP_016866828.1
6
1491
XM_017011340.1
XP_016866829.1
6
1543
XM_017011341.1
XP_016866830.1
4
1242
XM_017011342.1
XP_016866831.1
5
1383
XM_017011343.1
XP_016866832.1
6
1492
GenBank mRNA
AJ315122.1
-
1
299
AJ420549.1
-
2
369
AK027436.1
-
2
438
AK027558.1
-
3
486
AK027776.1
-
1
309
AL136829.1
-
5
697
BC008235.1
-
2
386
3868592
BI771613.1
-
2
436
CN388414.1
-
3
577
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
Wnt signaling pathway
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
protein autoubiquitination
protein K48-linked ubiquitination
positive regulation of canonical Wnt signaling pathway
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity
poly-ADP-D-ribose binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-97l6p:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline