Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243266.1 | NP_001230195.1 | 4 | 507 | ||
NM_024021.3 | NP_076926.2 | 5 | 653 | |||
NM_148975.2 | NP_683876.1 | 4 | 507 | |||
XM_017017909.1 | XP_016873398.1 | 4 | 520 | |||
GenBank mRNA | AB013102.1 | - | 4 | 348 | ||
AB022821.1 | - | 4 | 372 | |||
AF068288.1 | - | 4 | 441 | |||
AF237912.1 | - | 4 | 458 | |||
AF350500.1 | - | 3 | 411 | |||
AF354928.1 | - | 3 | 331 | |||
AK075081.1 | - | 5 | 538 | |||
AK292089.1 | - | 4 | 462 | |||
AK314033.1 | - | 4 | 350 | |||
BC020648.1 | - | 4 | 382 | 4720088 | ||
BG538851.1 | - | 4 | 508 | |||
CB046393.1 | - | 4 | 376 |