Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLP1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002062.4 | NP_002053.3 | 13 | 2149 | ||
NR_136563.1 | 14 | 1991 | ||||
GenBank mRNA | U01104.1 | - | 13 | 2126 | ||
U01156.1 | - | 14 | 1931 | |||
U01157.1 | - | 13 | 2085 |
Set Membership: |
G-protein coupled receptor |