Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000586.3 | NP_000577.2 | 1 | 147 | ||
XM_017008177.1 | XP_016863666.1 | 1 | 113 | |||
GenBank mRNA | A14844.1 | - | 1 | 102 | ||
AB451325.1 | - | 1 | 92 | |||
AB451454.1 | - | 1 | 92 | |||
AF228636.1 | - | 1 | 104 | |||
AY283686.1 | - | 1 | 32 | |||
AY523040.1 | - | 1 | 32 | |||
BC066254.1 | - | 1 | 110 | 6971690 | ||
BC066255.1 | - | 1 | 110 | 6971691 | ||
BC066256.1 | - | 1 | 110 | 6971693 | ||
BC066257.1 | - | 1 | 110 | 6971694 | ||
BC070338.1 | - | 1 | 139 | 30418817 | ||
DQ861285.1 | - | 1 | 94 | |||
S77834.1 | - | 1 | 139 | |||
S82692.1 | - | 1 | 386 | |||
U25676.1 | - | 1 | 148 | |||
V00564.1 | - | 1 | 139 | |||
X01586.1 | - | 1 | 199 |