Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEIS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002398.2 | NP_002389.1 | 2 | 681 | ||
XM_005264321.2 | XP_005264378.1 | 3 | 1004 | |||
XM_005264322.2 | XP_005264379.1 | 2 | 956 | |||
XM_005264323.2 | XP_005264380.1 | 3 | 1004 | |||
XM_017004141.1 | XP_016859630.1 | 3 | 1004 | |||
XM_017004142.1 | XP_016859631.1 | 2 | 956 | |||
XM_017004144.1 | XP_016859633.1 | 3 | 1004 | |||
XM_017004145.1 | XP_016859634.1 | 2 | 956 | |||
XM_017004146.1 | XP_016859635.1 | 2 | 956 | |||
GenBank mRNA | AI203963.1 | - | 2 | 345 | ||
AK297817.1 | - | 1 | 142 | |||
AK314949.1 | - | 2 | 431 | |||
AL832770.1 | - | 2 | 694 | |||
BC043503.1 | - | 2 | 676 | 5531644 | ||
U85707.1 | - | 2 | 289 |