Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005363.3 | NP_005354.1 | 3 | 670 | ||
NM_175868.2 | NP_787064.1 | 3 | 734 | |||
GenBank mRNA | AK289437.1 | - | 3 | 654 | ||
BC041599.1 | - | 3 | 643 | 5458512 | ||
BM015284.1 | - | 3 | 671 | |||
BU182516.1 | - | 3 | 684 | |||
BX640600.1 | - | 3 | 1031 | |||
D32076.1 | - | 1 | 528 | |||
EU161101.1 | - | 1 | 463 | |||
U10339.1 | - | 1 | 463 |
Gene Symbol: MAGEA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR541768.1 | - | 1 | 463 | ||
CR542020.1 | - | 1 | 463 |