Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270660.1 | NP_001257589.1 | 15 | 1488 | ||
NM_001270661.1 | NP_001257590.1 | 13 | 1317 | |||
NM_005885.3 | NP_005876.2 | 16 | 1632 | |||
XM_011513932.2 | XP_011512234.1 | 16 | 1390 | |||
XM_011513934.1 | XP_011512236.1 | 16 | 1593 | |||
XM_011513936.2 | XP_011512238.1 | 16 | 1593 | |||
XM_011513937.2 | XP_011512239.1 | 6 | 974 | |||
GenBank mRNA | AB011169.1 | - | 16 | 1555 | ||
AF009301.1 | - | 3 | 251 | |||
AK296585.1 | - | 13 | 1240 | |||
AK300034.1 | - | 15 | 1424 | |||
AK314686.1 | - | 5 | 485 | |||
BC046148.1 | - | 8 | 549 | |||
BC136461.1 | - | 16 | 1504 | 9020450 | ||
BC136462.1 | - | 16 | 1504 | 9020451 | ||
BC142679.1 | - | 16 | 1483 | 40148250 | ||
BC142694.1 | - | 16 | 1483 | 40148713 |