Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFGEF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020340.4 | NP_065073.3 | 12 | 1582 | ||
GenBank mRNA | AB033070.2 | - | 3 | 496 | ||
AB252196.1 | - | 12 | 1582 | |||
AF413080.1 | - | 1 | 190 | |||
AK124312.1 | - | 8 | 1020 | |||
BC137174.1 | - | 10 | 1217 | 9021171 |