Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077525.2 | NP_001070993.1 | 4 | 631 | ||
NM_001077526.2 | NP_001070994.1 | 4 | 631 | |||
NM_022485.4 | NP_071930.2 | 4 | 631 | |||
XM_005265400.1 | XP_005265457.1 | 4 | 604 | |||
XM_011534012.1 | XP_011532314.1 | 3 | 400 | |||
XM_011534013.1 | XP_011532315.1 | 3 | 356 | |||
XM_017007034.1 | XP_016862523.1 | 5 | 801 | |||
XM_017007035.1 | XP_016862524.1 | 5 | 801 | |||
XM_017007036.1 | XP_016862525.1 | 5 | 800 | |||
XM_017007037.1 | XP_016862526.1 | 5 | 801 | |||
XM_017007038.1 | XP_016862527.1 | 4 | 604 | |||
XM_017007039.1 | XP_016862528.1 | 5 | 800 | |||
XM_017007040.1 | XP_016862529.1 | 5 | 801 | |||
XM_017007041.1 | XP_016862530.1 | 3 | 400 | |||
XM_017007042.1 | XP_016862531.1 | 4 | 604 | |||
XM_017007043.1 | XP_016862532.1 | 3 | 400 | |||
GenBank mRNA | AF289603.1 | - | 4 | 594 | ||
AK026058.1 | - | 3 | 198 | |||
AK074792.1 | - | 4 | 602 | |||
AL713695.1 | - | 4 | 552 | |||
AY007098.1 | - | 1 | 39 | |||
BC001674.1 | - | 4 | 536 | 2961594 | ||
BC025952.1 | - | 4 | 581 | 4876556 | ||
BC035690.1 | - | 4 | 543 | 5755887 | ||
BC050626.1 | - | 4 | 554 | 4416416 | ||
DQ630520.1 | - | 4 | 480 |