Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303512.1 | NP_001290441.1 | 2 | 361 | ||
NM_001303513.1 | NP_001290442.1 | 2 | 361 | |||
NM_001303515.1 | NP_001290444.1 | 2 | 196 | |||
NM_001303516.1 | NP_001290445.1 | 2 | 196 | |||
NM_032512.3 | NP_115901.2 | 2 | 361 | |||
GenBank mRNA | AB103478.1 | - | 2 | 342 | ||
AK091444.1 | - | 2 | 87 | |||
AK123822.1 | - | 2 | 196 | |||
AK127016.1 | - | 2 | 361 | |||
AK309212.1 | - | 2 | 166 | |||
BC117277.1 | - | 2 | 153 | 40125828 | ||
BC143545.1 | - | 2 | 152 | 9052056 | ||
DA097500.1 | - | 2 | 182 |