Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZGRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018392.4 | NP_060862.3 | 25 | 5899 | ||
XM_005263115.3 | XP_005263172.1 | 25 | 5954 | |||
XM_011532091.2 | XP_011530393.1 | 24 | 5848 | |||
XM_011532092.2 | XP_011530394.1 | 24 | 5779 | |||
XM_011532093.2 | XP_011530395.1 | 24 | 5725 | |||
XM_011532094.2 | XP_011530396.1 | 23 | 5641 | |||
XM_011532097.2 | XP_011530399.1 | 18 | 3001 | |||
XM_011532098.2 | XP_011530400.1 | 16 | 2675 | |||
XM_011532099.2 | XP_011530401.1 | 15 | 2634 | |||
XM_017008369.1 | XP_016863858.1 | 24 | 5894 | |||
XM_017008370.1 | XP_016863859.1 | 23 | 5674 | |||
XM_017008371.1 | XP_016863860.1 | 22 | 5554 | |||
XM_017008372.1 | XP_016863861.1 | 22 | 4933 | |||
XM_017008373.1 | XP_016863862.1 | 21 | 4870 | |||
GenBank mRNA | AK090556.1 | - | 7 | 1534 | ||
AK126054.1 | - | 14 | 2283 | |||
AL137700.1 | - | 10 | 1346 | |||
BX647594.1 | - | 13 | 2254 |