Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145282.4 | NP_660325.4 | 3, 4 | 335 | ||
XM_005271903.1 | XP_005271960.1 | 3, 4 | 333 | |||
XM_006714544.1 | XP_006714607.1 | 3, 4 | 333 | |||
XM_006714546.1 | XP_006714609.1 | 3, 4 | 333 | |||
XM_006714550.1 | XP_006714613.1 | 3, 4 | 333 | |||
XM_006714552.2 | XP_006714615.1 | 3, 4 | 333 | |||
XM_011543196.1 | XP_011541498.1 | 3, 4 | 333 | |||
XM_011543199.1 | XP_011541501.1 | 3, 4 | 333 | |||
XM_017009086.1 | XP_016864575.1 | 3, 4 | 333 | |||
XM_017009087.1 | XP_016864576.1 | 3, 4 | 333 | |||
XM_017009088.1 | XP_016864577.1 | 3, 4 | 333 | |||
XM_017009089.1 | XP_016864578.1 | 3, 4 | 333 | |||
XM_017009090.1 | XP_016864579.1 | 6, 7 | 1445 | |||
XM_017009091.1 | XP_016864580.1 | 3, 4 | 333 | |||
XM_017009092.1 | XP_016864581.1 | 6, 7 | 1445 | |||
GenBank mRNA | AK126812.1 | - | 3, 4 | 335 | ||
BC025747.1 | - | 3, 4 | 284 | 5200129 | ||
BG700532.1 | - | 3, 4 | 319 | |||
DA137264.1 | - | 3, 4 | 333 |