Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145294.4 | NP_660337.3 | 39 | 5033 | ||
XM_017023023.1 | XP_016878512.1 | 40 | 5303 | |||
XM_017023024.1 | XP_016878513.1 | 22 | 2916 | |||
GenBank mRNA | AK092719.1 | - | 10 | 1749 | ||
AK093609.1 | - | 4 | 1433 | |||
AK131510.1 | - | 22 | 3339 | |||
BC065836.1 | - | 4 | 945 |