Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFESD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131065.1 | NP_001124537.1 | 4 | 1639 | ||
NM_001131066.1 | NP_001124538.1 | 4 | 484 | |||
NM_173362.3 | NP_775498.1 | 3 | 1541 | |||
XM_011543360.2 | XP_011541662.1 | 5 | 1527 | |||
XM_011543361.2 | XP_011541663.1 | 3 | 419 | |||
XM_017009413.1 | XP_016864902.1 | 4 | 651 | |||
GenBank mRNA | BC024023.1 | - | 2 | 258 | 4670524 | |
BC035110.2 | - | 3 | 1539 | 5262922 | ||
BC048979.1 | - | 3 | 1566 | 4838006 | ||
BI551847.1 | - | 4 | 487 | |||
BM263054.1 | - | 3 | 448 |