Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDLRAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174902.3 | NP_777562.1 | 2 | 223 | ||
GenBank mRNA | AK075546.1 | - | 2 | 142 | ||
AK299186.1 | - | 2 | 142 | |||
BC042754.1 | - | 2 | 126 | 4828259 | ||
BC136286.1 | - | 2 | 223 | 9020273 |