Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257359.1 | NP_001244288.1 | 6 | 1013 | ||
NM_174920.3 | NP_777580.1 | 6 | 1013 | |||
XM_011524490.2 | XP_011522792.1 | 3 | 199 | |||
XM_017024322.1 | XP_016879811.1 | 5 | 1397 | |||
XM_017024323.1 | XP_016879812.1 | 7 | 1180 | |||
XM_017024324.1 | XP_016879813.1 | 6 | 1868 | |||
GenBank mRNA | AK094209.1 | - | 6 | 944 | ||
AK095809.1 | - | 3 | 314 | |||
AK225767.1 | - | 6 | 944 | |||
AK291482.1 | - | 6 | 940 | |||
AY148482.1 | - | 6 | 934 | |||
BC029872.1 | - | 2 | 125 | |||
BC047081.1 | - | 5 | 667 | |||
BC136485.1 | - | 6 | 926 | 9020474 | ||
BC141822.1 | - | 6 | 953 | 40148162 | ||
BC142728.1 | - | 6 | 910 | 40148957 |