Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000365.5 | NP_000356.1 | 3 | 340 | ||
NM_001159287.1 | NP_001152759.1 | 3 | 449 | |||
NM_001258026.1 | NP_001244955.1 | 3 | 583 | |||
GenBank mRNA | AK129488.1 | - | 3 | 352 | ||
AK222638.1 | - | 3 | 329 | |||
AK298809.1 | - | 3 | 584 | |||
AK300661.1 | - | 3 | 228 | |||
AK313282.1 | - | 3 | 336 | |||
BC007086.1 | - | 3 | 336 | 4244564 | ||
BC007812.2 | - | 3 | 321 | 4298901 | ||
BC009329.2 | - | 3 | 327 | 4126160 | ||
BC011611.2 | - | 3 | 326 | 3956298 | ||
BC015100.1 | - | 3 | 341 | 3829453 | ||
BC017165.1 | - | 2 | 86 | |||
BC017917.1 | - | 3 | 332 | 4296431 | ||
BC018899.1 | - | 751 | ||||
BC070129.1 | - | 1 | 338 | 6725406 | ||
CR541702.1 | - | 3 | 302 | |||
EU794668.1 | - | 3 | 341 | |||
M10036.1 | - | 3 | 669 |