Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WWP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007013.3 | NP_008944.1 | 4 | 403 | ||
XM_005250760.3 | XP_005250817.1 | 3 | 364 | |||
XM_005250761.3 | XP_005250818.1 | 4 | 447 | |||
XM_011516802.2 | XP_011515104.1 | 3 | 164 | |||
XM_017012993.1 | XP_016868482.1 | 3 | 359 | |||
XM_017012994.1 | XP_016868483.1 | 3 | 354 | |||
XM_017012995.1 | XP_016868484.1 | 4 | 430 | |||
XM_017012996.1 | XP_016868485.1 | 3 | 348 | |||
XM_017012997.1 | XP_016868486.1 | 3 | 327 | |||
GenBank mRNA | AK292800.1 | - | 4 | 418 | ||
AL136739.1 | - | 2 | 133 | |||
AY043361.1 | - | 3 | 208 | |||
AY345857.1 | - | 4 | 437 | |||
BC036065.1 | - | 4 | 407 | 5296005 |