Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015336.2 | NP_056151.2 | 2 | 341 | ||
XM_005268750.4 | XP_005268807.1 | 2 | 181 | |||
XM_011538083.1 | XP_011536385.1 | 2 | 205 | |||
XM_017019091.1 | XP_016874580.1 | 2 | 181 | |||
XM_017019092.1 | XP_016874581.1 | 1 | 85 | |||
GenBank mRNA | AB023163.1 | - | 2 | 285 | ||
AB024494.1 | - | 2 | 828 | |||
AB097013.1 | - | 2 | 161 | |||
AB097035.1 | - | 2 | 161 | |||
AF049612.1 | - | 2 | 174 | |||
AF161412.1 | - | 2 | 173 | |||
AK292866.1 | - | 2 | 157 | |||
BC050324.1 | - | 2 | 300 | 5266097 | ||
DA227374.1 | - | 2 | 341 |