Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCHSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033449.2 | NP_258260.1 | 3 | 201 | ||
XM_005268524.4 | XP_005268581.1 | 3 | 234 | |||
XM_006714803.3 | XP_006714866.1 | 3 | 234 | |||
XM_011537698.2 | XP_011536000.1 | 3 | 234 | |||
XM_011537700.2 | XP_011536002.1 | 3 | 235 | |||
XM_011537701.2 | XP_011536003.1 | 3 | 235 | |||
XM_017010013.1 | XP_016865502.1 | 3 | 234 | |||
GenBank mRNA | AK000007.1 | - | 3 | 185 | ||
AY217346.1 | - | 3 | 201 | |||
BC047016.1 | - | 2 | 134 | 5215830 |