Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145282.4 | NP_660325.4 | 4, 5 | 604 | ||
XM_006714550.1 | XP_006714613.1 | 6, 7 | 994 | |||
XM_011543199.1 | XP_011541501.1 | 6, 7 | 990 | |||
XM_017009086.1 | XP_016864575.1 | 7, 8 | 1124 | |||
XM_017009090.1 | XP_016864579.1 | 10, 11 | 2236 | |||
GenBank mRNA | BC025747.1 | - | 4, 5 | 553 | 5200129 |