Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278505.1 | NP_001265434.1 | 5 | 429 | ||
NM_001322249.1 | NP_001309178.1 | 6 | 656 | |||
NM_174931.3 | NP_777591.3 | 7 | 698 | |||
XM_017003748.1 | XP_016859237.1 | 7 | 599 | |||
GenBank mRNA | AK095667.1 | - | 5 | 362 | ||
AK289504.1 | - | 5 | 344 | |||
AK294697.1 | - | 7 | 612 | |||
BC071798.1 | - | 5 | 361 | |||
BM285380.1 | - | 7 | 698 |