Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270660.1 | NP_001257589.1 | 18, 19 | 2019 | ||
NM_001270661.1 | NP_001257590.1 | 16, 17 | 1848 | |||
NM_005885.3 | NP_005876.2 | 19, 20 | 2163 | |||
XM_011513932.2 | XP_011512234.1 | 19, 20 | 1921 | |||
XM_011513934.1 | XP_011512236.1 | 19, 20 | 2124 | |||
XM_011513937.2 | XP_011512239.1 | 9, 10 | 1505 | |||
GenBank mRNA | AB011169.1 | - | 19, 20 | 2086 | ||
AF009301.1 | - | 6, 7 | 782 | |||
AK296585.1 | - | 16, 17 | 1771 | |||
AK300034.1 | - | 18, 19 | 1955 | |||
AK314686.1 | - | 8, 9 | 1016 | |||
BC046148.1 | - | 11, 12 | 1080 | |||
BC136461.1 | - | 19, 20 | 2035 | 9020450 | ||
BC136462.1 | - | 19, 20 | 2035 | 9020451 | ||
BC142679.1 | - | 19, 20 | 2014 | 40148250 | ||
BC142694.1 | - | 19, 20 | 2014 | 40148713 |