Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOBTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242359.1 | NP_001229288.1 | 7 | 1112 | ||
NM_014836.4 | NP_055651.1 | 6 | 1040 | |||
NR_024554.2 | 7 | 914 | ||||
NR_024555.1 | 7 | 878 | ||||
XM_006718084.1 | XP_006718147.1 | 6 | 848 | |||
XM_006718085.1 | XP_006718148.1 | 6 | 875 | |||
XM_011540424.1 | XP_011538726.1 | 7 | 906 | |||
XM_011540425.1 | XP_011538727.1 | 7 | 906 | |||
XM_011540426.1 | XP_011538728.1 | 7 | 906 | |||
XM_011540428.1 | XP_011538730.1 | 7 | 906 | |||
XM_017017000.1 | XP_016872489.1 | 6 | 1095 | |||
XM_017017001.1 | XP_016872490.1 | 6 | 1091 | |||
XM_017017002.1 | XP_016872491.1 | 6 | 1091 | |||
XM_017017003.1 | XP_016872492.1 | 8 | 981 | |||
XM_017017004.1 | XP_016872493.1 | 8 | 983 | |||
XM_017017005.1 | XP_016872494.1 | 7 | 888 | |||
XM_017017006.1 | XP_016872495.1 | 7 | 937 | |||
XM_017017007.1 | XP_016872496.1 | 5 | 987 | |||
XM_017017008.1 | XP_016872497.1 | 7 | 1148 | |||
GenBank mRNA | AB018283.3 | - | 6 | 931 | ||
AK075129.1 | - | 7 | 878 | |||
AK307630.1 | - | 5 | 610 | |||
BC032848.1 | - | 7 | 1089 | 4830471 | ||
BC041791.1 | - | 6 | 831 | 5297756 |