Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFGEF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020340.4 | NP_065073.3 | 24 | 4170 | ||
GenBank mRNA | AB033070.2 | - | 15 | 3084 | ||
AB252196.1 | - | 24 | 4170 | |||
AF413080.1 | - | 13 | 2778 | |||
BC137174.1 | - | 22 | 3805 | 9021171 |