Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF16B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199865.1 | NP_001186794.1 | 20, 21 | 3461 | ||
NM_001199866.1 | NP_001186795.1 | 21, 22 | 3614 | |||
NM_024704.4 | NP_078980.3 | 21, 22 | 3614 | |||
XM_005260750.3 | XP_005260807.1 | 22, 23 | 3626 | |||
XM_005260751.4 | XP_005260808.1 | 21, 22 | 3609 | |||
XM_005260752.3 | XP_005260809.1 | 21, 22 | 3473 | |||
XM_005260753.3 | XP_005260810.1 | 22, 23 | 3624 | |||
XM_005260754.3 | XP_005260811.1 | 22, 23 | 3624 | |||
XM_005260755.3 | XP_005260812.1 | 21, 22 | 3517 | |||
XM_006723588.3 | XP_006723651.1 | 22, 23 | 3624 | |||
XM_017027926.1 | XP_016883415.1 | 21, 22 | 3609 | |||
GenBank mRNA | AB046810.2 | - | 21, 22 | 3531 | ||
AK026698.1 | - | 2, 3 | 203 | |||
AY044654.1 | - | 1, 2 | 116 | |||
AY166853.1 | - | 21, 22 | 3613 | |||
BC034984.1 | - | 3, 4 | 290 | |||
BC050577.1 | - | 20, 21 | 3461 | |||
BC110317.1 | - | 3, 4 | 710 | |||
BC150261.1 | - | 21, 22 | 3531 | 9007204 | ||
BX647572.1 | - | 21, 22 | 3533 | |||
BX648426.1 | - | 7, 8 | 1319 |