Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBA1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303114.1 | NP_001290043.1 | 4 | 1573 | ||
NM_001303115.1 | NP_001290044.1 | 6 | 1806 | |||
NM_001303116.1 | NP_001290045.1 | 5 | 1688 | |||
NM_001303117.1 | NP_001290046.1 | 5 | 1691 | |||
NM_032704.4 | NP_116093.1 | 4 | 1518 | |||
GenBank mRNA | AK223024.1 | - | 4 | 1435 | ||
AK293589.1 | - | 4 | 1573 | |||
BC004949.1 | - | 4 | 1415 | 3617714 | ||
BC005946.1 | - | 4 | 1435 | 4134187 | ||
BC011790.1 | - | 4 | 1416 | 4025017 | ||
BC019298.2 | - | 4 | 1397 | 2905506 | ||
BC021088.1 | - | 4 | 1396 | 3543481 | ||
BC033064.1 | - | 5 | 933 | 3926854 | ||
BC051297.1 | - | 4 | 1411 | 3832363 | ||
BC063036.1 | - | 4 | 1457 | 4397636 |